Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
651101 is a mixture of monoclonal antibodies that target Plakophilin 2 Multiepitope.
Specificity: plakophilin 2, a member of the arm-repeat family of proteins which can occur in at least 2 splice forms (2a and 2b). Plakophilin 2 assembles with other proteins to form the desmosomal plaque in simple and glandular epithelia, the basal layer of certain stratified epithelia, all layers of some stratified epithelia, and in nonepithelial desmosome-possessing tissues such as myocardium, Purkinje fibers, and dendritic reticulum of lymph node follicles. In all these cells, including a variety of cell types devoid of desmosomes, plakophilin 2 is highly enriched in the karyoplasm. Polypeptide reacting: Plakophilin 2, MW 100 kD (SDS-PAGE); MW calculated from aa sequence: 92,750 (pI 9.33) and 97,410 (pI 9.38)Tumors specifically detected: Positive staining was obtained with several carcinomas derived from simple-type epithelia (e.g. hepatomas, colon carcinomas) and some adenocarcinomas derived from stratified epithelia (oesophagus carcinoma).
Plakophilins 1, 2, 3 and 4 (PKP1-4) influence development and participate in linking cadherins to cytoskeletal intermediate filaments. Plakophilins 1-4 contain arm-repeat (armadillo) domains and localize to nuclei and cell desmosomes (cell-cell junctions found in suprabasal layers of stratifying epithelia that undergo mechanical stress). Plakophilin 1 mediates increases in desmosomal protein content, desmosome assembly and regulation of cell migration. Plakophilin 2 is important for desmosome assembly and is an essential morphogenic factor and architectural component of the heart. Plakophilin 4 is a component of desmosomal adhesion plaques that regulates junctional plaque organization and cadherin function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Plakophilin-2
Gene Aliases: ARVD9; PKP2
UniProt ID: (Human) B8QFA1, (Rat) F1M7L9
Entrez Gene ID: (Human) 5318, (Rat) 287925
Molecular Function:
intermediate filament binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support