Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Progen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Plakophilin 3 is seen in most desmosomes and partly dispersed in cytoplasm under various fixation conditions (FA, MeAc long, MeAc short):Positive tissues and culture cells:stratified squamous epithelia: palmar, epidermis, scalp skinstratified epithelia: tongue, vagina, esophagustransitional epithelia: bladdernon-stratified epithelia: colon, duodenum, pancreas, several glandsnon-epithelial: reticulum cells of lymph node follicleHaCaT, MCF7, HeLa, HT-29, CaCo 2, Caski, BMGE, MDCK and otherNegative tissues and cell lines:liver, heart, skeletal muscle, endothelia (vessels)PLC, RD, SV80, HL60, Raji, Jok1, Glioma, K562, 3T3L1, A6, PtK2.
Plakophilin-3 is an Armadillo-like protein present in nuclei and desmosomes of epithelial cells. The expression pattern of this protein seems to be largely restricted to epithelial cell types. Plakophilin-3 can be detected along cell borders in a punctuate staining pattern typical for desmosomal proteins. In addition to the desmosomal immunolocalization, immunostaining was observed as bright nuclear speckles. Thus, like plakophilin-1 and-2, plakophilin-3 displays a dual intracellular localization in the desmosomal plaque and in the cell nucleus, and therefore is probably involved in signal transduction pathways between the plasma membrane and the nucleus. The human protein has a predicted molecular mass of ~87 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: plakophilin 3b; Plakophilin-3
Gene Aliases: 2310056L12Rik; PKP3
UniProt ID: (Human) Q9Y446, (Mouse) Q9QY23
Entrez Gene ID: (Human) 11187, (Mouse) 56460, (Rat) 293619
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support