FIGURE: 1 / 1
Immunogen sequence: YLQPAGDMSK SAVDGTAENP FLDNPDAFYY FPDSGFFCEE NSSDDDVILK EEFRGVIIKQ GCLLKQGHRR KNWKVRKFIL REDPAYLHYY DPAGAEDPLG AIHLRGCVVT SVESNSNGRK SEEENLFEII TADEVHYFLQ AATPKERTEW IRAIQMASRT GK; Positive Samples: THP-1, U-937, Raji
Major protein kinase C substrate of platelets.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ27168; p47; Platelet 47 kDa protein; Pleckstrin
Gene Aliases: P47; PLEK
UniProt ID: (Human) P08567
Entrez Gene ID: (Human) 5341
Molecular Function:
cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support