Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The antibody detects a 220 kDa* protein corresponding to the apparent molecular mass of Plexin-A1 on SDS-PAGE immunoblots of neonatal mouse brain, and detects a recombinant mouse Plexin A1 protein containing the extracellular region.
Coreceptor for SEMA3A, SEMA3C, SEMA3F and SEMA6D. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Plex 1; plexin 1; plexin A1; plexin-1; Plexin-A1; Semaphorin receptor NOV
Gene Aliases: 2600013D04Rik; Kiaa4053; mKIAA4053; NOV; NOVP; PlexA1; PLEXIN-A1; PLXN1; PLXNA1; RGD1560871
UniProt ID: (Human) Q9UIW2, (Mouse) P70206
Entrez Gene ID: (Human) 5361, (Rat) 362398, (Mouse) 18844
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support