Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C, protected from the light, for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 X g 5 min).
Receptor for SEMA7A, for smallpox semaphorin A39R, vaccinia virus semaphorin A39R and for herpesvirus Sema protein. Binding of semaphorins triggers cellular responses leading to the rearrangement of the cytoskeleton and to secretion of IL6 and IL8. {ECO:0000250, ECO:0000269; PubMed:20727575}.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD232; CD232 antigen; plexin (semaphorin receptor); Plexin-C1; receptor for viral semaphorin protein; receptor for virally-encoded semaphorin; VESPRplexin-C1; viral-encoded semaphorin protein receptor; Virus-encoded semaphorin protein receptor
Gene Aliases: 2510048K12Rik; AW742158; CD232; PLXN-C1; PLXNC1; VESPR
UniProt ID: (Human) O60486, (Mouse) Q9QZC2
Entrez Gene ID: (Human) 10154, (Rat) 362873, (Mouse) 54712
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support