Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 3
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Guinea pig_10141, Panda, Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon and Little brown bat
Mitochondrial creatine kinase (MtCK) is responsible for the transfer of high energy phosphate from mitochondria to the cytosolic carrier, creatine. It belongs to the creatine kinase isoenzyme family. It exists as two isoenzymes, sarcomeric MtCK and ubiquitous MtCK, encoded by separate genes. Mitochondrial creatine kinase occurs in two different oligomeric forms: dimers and octamers, in contrast to the exclusively dimeric cytosolic creatine kinase isoenzymes. Sarcomeric mitochondrial creatine kinase has 80% homology with the coding exons of ubiquitous mitochondrial creatine kinase. This gene contains sequences homologous to several motifs that are shared among some nuclear genes encoding mitochondrial proteins and thus may be essential for the coordinated activation of these genes during mitochondrial biogenesis. Three transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arsenic transactivated protein; Arsenic-transactivated protein; AsTP; CHRAC 17; CHRAC-17; Chromatin accessibility complex 17 kDa protein; DNA polymerase epsilon p17 subunit; DNA polymerase epsilon subunit 3; DNA polymerase epsilon subunit p17; DNA polymerase epsilon, subunit 3; DNA polymerase II subunit 3; histone fold protein CHRAC17; huCHRAC17; NF-YB-like protein; polymerase (DNA directed), epsilon 3 (p17 subunit); polymerase (DNA directed), epsilon 3, accessory subunit; polymerase (DNA) epsilon 3, accessory subunit; YB-like protein 1; YBL1
Gene Aliases: 1810034K18Rik; CHARAC17; CHRAC17; p17; POLE3; YBL1
UniProt ID: (Human) Q9NRF9, (Mouse) Q9JKP7
Entrez Gene ID: (Human) 54107, (Mouse) 59001
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support