Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alzheimer's disease (AD) patients with an inherited form of the disease carry mutations in the presenilin proteins (PSEN1; PSEN2) or the amyloid precursor protein (APP). These disease-linked mutations result in increased production of the longer form of amyloid-beta (main component of amyloid deposits found in AD brains). Presenilins are postulated to regulate APP processing through their effects on gamma-secretase, an endoprotease complex that catalyzes the intramembrane cleavage of integral membrane proteins such as Notch receptors and APP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AD3LP; AD5; Alzheimer disease 4; E5-1; Presenilin-1; Presenilin-2; Protein S182; PS-1; PS-2; STM-2
Gene Aliases: AD3; Ad3h; AD3L; AD4; Ad4h; ALG-3; Alg3; CMD1V; FAD; PS-1; PS-2; PS1; PS2; PSEN1; PSEN2; PSNL1; PSNL2; S182; STM2
UniProt ID: (Human) P49768, (Human) P49810, (Rat) P97887, (Rat) O88777, (Mouse) Q61144
Entrez Gene ID: (Human) 5663, (Human) 5664, (Rat) 29192, (Mouse) 19164, (Rat) 81751, (Mouse) 19165
Molecular Function: aspartic protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support