Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
BTE 003-02-02 is specific human PIIANP.
BTE 003-02-02 has been successfully used ELISA, and Immunohistochemistry procedures.
Epitope is located at the amino-terminal part of procollagen type IIA.
NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Collagen type II is the most abundant protein of cartilage matrix. Two isoforms exist and collagen type IIA is involved in early development of cartilage in embryos. The collagen is produced from type II procollagen when two teloterminal propeptides are digested from the procursor and released into the circulation. Procollagen type II N-terminal propeptide (PIIANP) is considered a marker of cartilage synthesis and the normal concentration in serum is approximately 30 ng/mL.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-1 type II collagen; arthroophthalmopathy, progressive (Stickler syndrome); cartilage collagen; chondrocalcin; Collagen alpha-1(II) chain; collagen II, alpha-1 polypeptide; collagen, type II, alpha 1
Gene Aliases: ANFH; AOM; COL11A3; COL2A1; SEDC; STL1
UniProt ID: (Human) P02458
Entrez Gene ID: (Human) 1280
Molecular Function: extracellular matrix structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support