Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MARFLTLCT WLLLLGPGLL ATVRAECSQD CATCSYRLVR PADINFLACV MECEGKLPSL KIWETCKELL QLSKPELPQD GTSTLRENSK PEESHLLAKR YGGFMKRYGG FMKKMDELYP MEPEEEANGS EILAKRYGGF MKKDAEEDDS LANSSDLLKE LLETGDNRER SHHQDGSDNE EEVSKRYGGF MRGLKRSPQL EDEAKELQKR YGGFMRRVGR PEWWMDYQKR YGGFLKRFAE ALPSDEEGES YSKEVPEMEK RYGGFMRF (1-267 aa encoded by BC032505)
neuropeptide hormone; plays a role in the responses to stress and pain perception.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: enkephalin A; preproenkephalin; Proenkephalin-A
Gene Aliases: PE; PENK; PENK-A
UniProt ID: (Human) P01210
Entrez Gene ID: (Human) 5179
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support