Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
OPA1-15292 has been successfully used in immunohistochemistry procedures. By immunohistochemistry, OPA1-15292 detects the prokineticin receptor 2 (GPR73B) in formalin fixed, paraffin embedded human tissues.
Prokineticins are secreted proteins that can promote angiogenesis and induce strong gastrointestinal smooth muscle contraction. GPR73B is an integral membrane protein and G protein-coupled receptor for prokineticins. The encoded protein is similar in sequence to GPR73, another G protein-coupled receptor for prokineticins. GPCR73B expression has been reported in brain, ovary, salivary gland, small intestine (ileocecum), testis, and thyroid. No human ESTs have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 73-like 1; G-protein coupled receptor 73-like 1; G-protein coupled receptor I5E; GPCR73B; GPR73b; GPRg2; OTTHUMP00000030192; PK-R2; Prokineticin receptor 2
Gene Aliases: dJ680N4.3; GPR73b; GPR73L1; GPRg2; HH3; KAL3; PKR2; PROKR2
UniProt ID: (Human) Q8NFJ6
Entrez Gene ID: (Human) 128674
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support