Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset (100%); Elephant, Horse, Opossum (93%); Gorilla, Panda, Dog, Bat, Bovine, Rabbit, Platypus (87%).
Proteinase K treatment is required for antigen retrieval.
Prokineticins are secreted proteins that can promote angiogenesis and induce strong gastrointestinal smooth muscle contraction. GPR73B is an integral membrane protein and G protein-coupled receptor for prokineticins. The encoded protein is similar in sequence to GPR73, another G protein-coupled receptor for prokineticins. GPCR73B expression has been reported in brain, ovary, salivary gland, small intestine (ileocecum), testis, and thyroid. No human ESTs have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 73-like 1; G-protein coupled receptor 73-like 1; G-protein coupled receptor I5E; GPCR73B; GPR73b; GPRg2; OTTHUMP00000030192; PK-R2; Prokineticin receptor 2
Gene Aliases: dJ680N4.3; GPR73b; GPR73L1; GPRg2; HH3; KAL3; PKR2; PROKR2
UniProt ID: (Human) Q8NFJ6
Entrez Gene ID: (Human) 128674
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support