Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Plays an important role in the regulation of interdigestive gastrointestinal motility and indirectly causes rhythmic contraction of duodenal and colonic smooth muscle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: prepromotilin; Promotilin
Gene Aliases: MLN
UniProt ID: (Human) P12872
Entrez Gene ID: (Human) 4295, (Rat) 100126231
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support