Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Exalpha
Positive control: Porcine brain lysate.
This polyclonal antibody was raised in Rabbit against PTGER3 / EP3. This antibody has been conjugated with Unconjugated. This antibody is useful for testing in the following assays: IHC-P. The specificity of this antibody has been described as follows: Human PTGER3 / EP3. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EP2 receptor; PGE receptor EP2 subtype; PGE receptor EP3 subtype; PGE receptor, EP2 subtype; PGE receptor, EP3 subtype; PGE2 receptor EP2 subtype; PGE2 receptor EP3 subtype; PGE2-R; prostaglandin E receotor EP3 subtype 3 isoform; prostaglandin E receptor 2 (EP2 subtype); prostaglandin E receptor 2 (subtype EP2), 53kD; prostaglandin E receptor 2 (subtype EP2), 53kDa; prostaglandin E receptor 3 (subtype EP3); prostaglandin E receptor EP2 subtype; prostaglandin E receptor EP3 subtype; prostaglandin E receptor subtype EP2; Prostaglandin E2 receptor EP2 subtype; Prostaglandin E2 receptor EP3 subtype; prostaglandin E2 receptor type 2; prostaglandin receptor (PGE-2); Prostanoid EP2 receptor; Prostanoid EP3 receptor
Gene Aliases: EP2; EP3; EP3-I; EP3-II; EP3-III; EP3-IV; EP3-VI; EP3e; PGE2-R; Pgerep3; Ptger-ep2; PTGER2; PTGER3; Ptgerep2; Ptgerep3; Rep3; rEP3a; rEP3b
UniProt ID: (Human) P43116, (Human) P43115, (Rat) Q62928, (Mouse) Q62053, (Rat) P34980
Entrez Gene ID: (Human) 5732, (Human) 5733, (Rat) 81752, (Mouse) 19217, (Rat) 24929, (Mouse) 19218
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support