Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive test controls include: Mouse kidney. The target is usually found in the following locations: Cell membrane, Secreted, Single-pass membrane protein, extracellular space, extracellular space.
Immunogen sequence: TCNCLYNIDA KPEEPHFVQE DMVCAGYVEG GKDACQGDSG GPLSCPVEGL WYLTGIVSWG DACGARNRPG VYTLASSYAS WIQSKVTELQ PRVVPQTQES QPDSNLCGSH LAFSSAPAQG LLRPILFLPL GLALGLLSPW LSEH
This gene encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is highly expressed in prostate epithelia and is one of several proteolytic enzymes found in seminal fluid. The proprotein is cleaved to produce a light chain and a heavy chain which are associated by a disulfide bond. It is active on peptide linkages involving the carboxyl group of lysine or arginine.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAP1; channel activating protease 1; Channel-activating protease 1; Prostasin; protease, serine, 8 (prostasin); Serine protease 8
Gene Aliases: 2410039E18Rik; AI313909; C79772; CAP1; fr; mCAP1; PROSTASIN; PRSS8
UniProt ID: (Human) Q16651, (Rat) Q9ES87
Entrez Gene ID: (Human) 5652, (Mouse) 76560, (Rat) 192107
Molecular Function: serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support