Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detects a primarily full length APC at 300 kDa* in human Hct116 cells, mouse brain, and in rat PC12 cells. In immunocytochemistry, the antibody detects in APC in clusters at the plasma membrane and at the ends of microtubules.
This gene encodes a tumor suppressor protein that acts as an antagonist of the Wnt signaling pathway. It is also involved in other processes including cell migration and adhesion, transcriptional activation, and apoptosis. Defects in this gene cause familial adenomatous polyposis (FAP), an autosomal dominant pre-malignant disease that usually progresses to malignancy. Disease-associated mutations tend to be clustered in a small region designated the mutation cluster region (MCR) and result in a truncated protein product.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adenomatosis polyposis coli; adenomatosis polyposis coli tumor suppressor; Adenomatous polyposis coli protein; Deleted in polyposis 2.5; FPC; multiple intestinal neoplasia; Protein APC; protein phosphatase 1, regulatory subunit 46; truncated adenomatosis polyposis coli; WNT signaling pathway regulator
Gene Aliases: AI047805; APC; AU020952; AW124434; BTPS2; CC1; DP2; DP2.5; DP3; GS; mAPC; Min; PPP1R46; RATAPC
UniProt ID: (Human) P25054, (Rat) P70478
Entrez Gene ID: (Human) 324, (Mouse) 11789, (Rat) 24205
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support