Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance of the antiserum. Diluted antiserum should be stored at 4 C, not refrozen, and preferably used the same day.
In immunoelectrophoresis use 2 µL serum, plasma or equivalent against 120 µL antiserum. In double radial immunodiffusion (Ouchterlony) use a rosette arrangement with 10 µL antiserum in a 3 mm diameter centre well and 2 µL serum samples (neat and serially diluted) in 2 mm diameter peripheral wells. In single radial immunodiffusion and electroimmunodiffusion use 0.5 to 1.0 percent antiserum in the agar gel.
The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase dephosphorylates, and negatively regulates the activities of, MAP kinases and MAP kinase kinases. It has been shown to inhibit the activation of p38 and JNK kinase cascades induced by environmental stresses. This phosphatase can also dephosphorylate cyclin-dependent kinases, and thus may be involved in cell cycle control. Overexpression of this phosphatase is reported to activate the expression of the tumor suppressor gene TP53/p53, which leads to G2/M cell cycle arrest and apoptosis. Three alternatively spliced transcript variants encoding distinct isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ42306; IA; MGC9201; Mpp alpha; PP2C-alpha; PP2C-beta; Protein phosphatase 1A; protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform; protein phosphatase 1A, magnesium dependent, alpha isoform; Protein phosphatase 1B; protein phosphatase 1B (formerly 2C), magnesium-dependent, beta isoform; protein phosphatase 1B, magnesium dependent, beta isoform; Protein phosphatase 2C isoform alpha; Protein phosphatase 2C isoform beta; protein phosphatase 2C-like protein; Protein phosphatase IA; Protein phosphatase type 1A (formely 2C) Mg-dependent alpha isoform; Protein phosphatase type 1A (formely 2C), Mg-dependent, alpha isoform; Protein phosphatase type 1B (formely 2C) Mg-dependent beta isoform; Protein phosphatase type 1B (formely 2C), Mg-dependent, beta isoform; protein phosphatase, Mg2+/Mn2+ dependent, 1A; protein phosphatase, Mg2+/Mn2+ dependent, 1B; Ser/Thr protein phosphatase type 2C beta 2 isoform
Gene Aliases: 2310003C21Rik; 2900017D14Rik; AI427932; AU017636; MMPa-2; MPPa-1; PP2C-ALPHA; PP2C-beta; PP2C-beta-X; Pp2c1; Pp2c2; PP2CA; PP2Calpha; PP2CB; PP2CBETA; PPC2BETAX; PPM1A; PPM1B; PPPM1A; Pppm1b
UniProt ID: (Human) P35813, (Bovine) O62829, (Bovine) O62830, (Human) O75688, (Mouse) P49443, (Rat) P20650, (Mouse) P36993, (Rat) P35815
Entrez Gene ID: (Human) 5494, (Bovine) 281994, (Bovine) 281995, (Human) 5495, (Mouse) 19042, (Rat) 24666, (Mouse) 19043, (Rat) 24667
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support