Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance of the antiserum. Diluted antiserum should be stored at 4 C, not refrozen, and preferably used the same day.
In immunoelectrophoresis use 2 µL serum, plasma or equivalent against 120 µL antiserum. In double radial immunodiffusion (Ouchterlony) use a rosette arrangement with 10 µL antiserum in a 3 mm diameter centre well and 2 µL serum samples (neat and serially diluted) in 2 mm diameter peripheral wells. In single radial immunodiffusion and electroimmunodiffusion use 0.5 to 1.0 percent antiserum in the agar gel.
Distinct DNA repair pathways minimize the consequences of mutagenic events. Reactive oxygen species (ROS) are highly reactive atoms with an unpaired electron that are conducive to double-strand DNA breaking events. Artemis, named after the Greek goddess for the protection of children, is one of the major proteins contributing to the preservation of double-strand breaks in DNA by cutting away the damaged parts of the DNA, which allows the strands to rejoin. Artemis is a single-strand-specific 5' to 3' exonuclease that forms a complex with the 469 kDa DNA-dependent protein kinase (DNA-PKcs). DNA-PKcs phosphorylates Artemis, and Artemis acquires endonucleolytic activity on 5' and 3' overhangs and hairpins. These activities are essential for V(D)J recombination and for the 5' and 3' overhang processing in nonhomologous DNA end joining.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ANKRD33; FLJ34236; PP-V; PP6C; protein phosphatase 6, catalytic subunit; Protein phosphatase V; serine/threonine protein phosphatase catalytic subunit; Serine/threonine-protein phosphatase 6 catalytic subunit; UNQ5837
Gene Aliases: 2310003C10Rik; PP6; PP6C; PPP6; PPP6C; Ppv
UniProt ID: (Human) O00743, (Rat) Q64620, (Mouse) Q9CQR6
Entrez Gene ID: (Bovine) 511058, (Human) 5537, (Rat) 171121, (Mouse) 67857
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support