View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
The antibody reacts with bacteria in suspension and is therefore useful for detection of Proteus spp. in bacterial suspensions. Various systems can be used with this antibody to detect the presence of Proteus spp.. The antibody reacts with approximately 50% of the Proteus spp. isolates tested.