Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-43039 detects Prothrombin in human samples.
PA1-43039 has successfully been used in Western blot and ELISA applications.
PA1-43039 was derived from human prothrombin.
Prothrombin or Coagulation factor II, is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coagulation factor II; coagulation factor II (thrombin); prepro-coagulation factor II; Prothrombin; prothrombin B-chain; serine protease
Gene Aliases: F2; PT; RPRGL2; THPH1
UniProt ID: (Human) P00734
Entrez Gene ID: (Human) 2147
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support