Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
QED Bioscience
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 3
The protocadherin gamma gene cluster is one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3' RACE product; cadherin 20; cadherin 21; cadherin ME3; cadherin ME6; Cadherin-20; Cadherin-21; cadherin-like 2; fibroblast cadherin FIB2; fibroblast cadherin FIB3; Fibroblast cadherin-2; Fibroblast cadherin-3; Pan-Gamma-Protocadherin-A; PC-43; PCDH gamma A; PCDH-gamma-A; PCDH-gamma-A1; PCDH-gamma-A10; PCDH-gamma-A11; PCDH-gamma-A12; PCDH-gamma-A2; PCDH-gamma-A3; PCDH-gamma-A4; PCDH-gamma-A5; PCDH-gamma-A6; PCDH-gamma-A7; PCDH-gamma-A8; PCDH-gamma-A9; PCDH-gamma-B1; PCDH-gamma-B2; PCDH-gamma-B3; PCDH-gamma-B4; PCDH-gamma-B5; PCDH-gamma-B6; PCDH-gamma-B7; PCDH-gamma-C3; PCDH-gamma-C4; PCDH-gamma-C5; PCDHGA; pcdhga4; protocadherin 13; protocadherin 2; protocadherin 43; protocadherin gamma a1; protocadherin gamma A12; protocadherin gamma a2; protocadherin gamma B2; protocadherin gamma B6; protocadherin gamma b7; protocadherin gamma C-VI; protocadherin gamma c5; protocadherin gamma subfamily A, 10; protocadherin gamma subfamily A, 12; protocadherin gamma subfamily A, 3; protocadherin gamma subfamily A, 4; protocadherin gamma subfamily B, 1, isoform 2; protocadherin gamma subfamily B, 5; protocadherin gamma subfamily B, 7; protocadherin gamma subfamily C, 3; Protocadherin gamma-A1; Protocadherin gamma-A10; Protocadherin gamma-A11; Protocadherin gamma-A12; Protocadherin gamma-A2; Protocadherin gamma-A3; Protocadherin gamma-A4; Protocadherin gamma-A5; Protocadherin gamma-A6; protocadherin gamma-A6-like; Protocadherin gamma-A7; Protocadherin gamma-A8; Protocadherin gamma-A9; Protocadherin gamma-B1; Protocadherin gamma-B2; Protocadherin gamma-B3; Protocadherin gamma-B4; protocadherin gamma-B4-like; Protocadherin gamma-B5; Protocadherin gamma-B6; protocadherin gamma-B6-like; Protocadherin gamma-B7; Protocadherin gamma-C3; Protocadherin gamma-C4; Protocadherin gamma-C5; Protocadherin-2; Protocadherin-43
Gene Aliases: CDH-GAMMA-A5; CDH20; CDH21; FIB2; FIB3; KIAA0327; KIAA0588; ME3; ME6; mKIAA0588; mKIAA4054; pc2c; PC43; PCDH-GAMMA-A1; PCDH-GAMMA-A10; PCDH-GAMMA-A11; PCDH-GAMMA-A12; PCDH-GAMMA-A2; PCDH-GAMMA-A3; PCDH-GAMMA-A4; PCDH-GAMMA-A5; PCDH-GAMMA-A6; PCDH-GAMMA-A7; PCDH-GAMMA-A8; PCDH-GAMMA-A9; PCDH-GAMMA-B1; PCDH-GAMMA-B2; PCDH-GAMMA-B3; PCDH-GAMMA-B4; PCDH-GAMMA-B5; PCDH-GAMMA-B6; PCDH-GAMMA-B7; PCDH-GAMMA-C3; PCDH-GAMMA-C4; PCDH-GAMMA-C5; Pcdh13; PCDH2; PCDHGA1; PCDHGA10; Pcdhga10temp; PCDHGA11; PCDHGA12; PCDHGA2; PCDHGA3; PCDHGA4; PCDHGA5; PCDHGA6; PCDHGA7; PCDHGA8; PCDHGA9; PCDHGB1; PCDHGB2; PCDHGB3; PCDHGB4; PCDHGB5; PCDHGB6; PCDHGB7; PCDHGC3; PCDHGC4; PCDHGC5; UNQ371/PRO707
UniProt ID: (Human) Q9Y5H4, (Human) Q9Y5H3, (Human) Q9Y5H2, (Human) O60330, (Human) Q9Y5H1, (Human) Q9Y5H0, (Human) Q9Y5G9, (Human) Q9Y5G8, (Human) Q9Y5G7, (Human) Q9Y5G6, (Human) Q9Y5G5, (Human) Q9Y5G4, (Human) Q9Y5G3, (Human) Q9Y5G2, (Human) Q9Y5G1, (Human) O15099, (Human) Q9Y5G0, (Human) Q9Y5F9, (Human) Q9Y5F8, (Human) Q9UN70, (Human) Q9Y5F7, (Human) Q9Y5F6, (Mouse) Q91XY4
Entrez Gene ID: (Rat) 102552497, (Rat) 102552705, (Rat) 102552796, (Human) 56114, (Human) 56106, (Human) 56105, (Human) 26025, (Human) 56113, (Human) 56112, (Human) 56111, (Human) 56110, (Human) 56109, (Human) 56108, (Human) 9708, (Human) 56107, (Human) 56104, (Human) 56103, (Human) 56102, (Human) 8641, (Human) 56101, (Human) 56100, (Human) 56099, (Human) 5098, (Human) 56098, (Human) 56097, (Rat) 553129, (Mouse) 93709, (Rat) 498849, (Mouse) 93722, (Rat) 252897, (Mouse) 93723, (Mouse) 93724, (Rat) 498850, (Rat) 498846, (Mouse) 93710, (Mouse) 93711, (Rat) 498847, (Mouse) 93712, (Rat) 252894, (Rat) 291637, (Mouse) 93713, (Mouse) 93714, (Mouse) 93715, (Rat) 291635, (Rat) 364843, (Mouse) 93716, (Rat) 252895, (Mouse) 93717, (Mouse) 93699, (Mouse) 93700, (Mouse) 93701, (Mouse) 93702, (Rat) 680448, (Mouse) 93703, (Mouse) 93704, (Rat) 364844, (Mouse) 93706, (Rat) 116782, (Mouse) 93707, (Mouse) 93708, (Rat) 641525
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support