Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: SRHLAAKMAS TPHPPGARGT SQLHGMDLLV LLDLIGAPNP TFPNFFPNSA
Sequence homology: Cow: 86%; Dog: 79%; Guinea Pig: 93%; Horse: 79%; Human: 100%; Mouse: 79%; Rabbit: 93%; Rat: 100%; Sheep: 86%
GPIP137 regulates the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation/migration in multiple cell types. It binds directly and selectively to MYC and CCND2 RNAs. In neuronal cells, GPIP137 binds to several mRNAs associated with RNA granules including BDNF, CAMK2A, CREB1, MAP2, NTRK2, mRNAs, GRIN1, and KPNB1 mRNAs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC; Glutaminyl cyclase; Glutaminyl-peptide cyclotransferase; Glutaminyl-tRNA cyclotransferase; Glutamyl cyclase; QC
Gene Aliases: GCT; QC; QPCT; sQC
UniProt ID: (Human) Q16769
Entrez Gene ID: (Human) 25797
Molecular Function: transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support