Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: GIPKEYLVP ELSSEVQSLW SKAADLFESE GAKVIEVSLP HTSYSIVCYH VLCTSEVASN MARFDGLQYG HRCDIDVSTE AMYAATRREG FNDVVRGRIL SGNFFLLKEN YENYFVKAQK VRRLIANDFV NAFNSGVDVL LTPTTLSEAV PYLEFIKEDN RTRSAQDDIF TQAVNMAGLP AVSIPVALSN QGLPIGLQFI GRAFCDQQLL TVAKWFEKQV QFPVIQLQEL MDDCSAVLEN EKLASVSLKQ (280-528 aa encoded by BC014389)
Allows the formation of correctly charged Gln-tRNA(Gln) through the transamidation of misacylated Glu-tRNA(Gln) in the mitochondria. The reaction takes place in the presence of glutamine and ATP through an activated gamma-phospho-Glu- tRNA(Gln).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glu-AdT subunit A; glu-AdT subunit A {ECO:0000255|HAMAP-Rule:MF_03150}; Glutaminyl-tRNA synthase-like protein 1; glutaminyl-tRNA synthase-like protein 1 {ECO:0000255|HAMAP-Rule:MF_03150}; glutamyl-tRNA(Gln) amidotransferase subunit A homolog; Glutamyl-tRNA(Gln) amidotransferase subunit A, mitochondrial; glutamyl-tRNA(Gln) amidotransferase subunit A, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03150}; glutamyl-tRNA(Gln) amidotransferase, subunit A; QRSL1
Gene Aliases: 2700038P16Rik; C80053; GatA; QRSL1
UniProt ID: (Human) Q9H0R6, (Rat) Q5FWT5, (Mouse) Q9CZN8
Entrez Gene ID: (Human) 55278, (Rat) 309911, (Mouse) 76563
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support