Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKLSLTKVVN GCRLGKIKNL GKTGDHTMDI PGCLLYTKTG SAPHLTHHTL HNIHGVPAMA QLTLSSLAEH HEVLTEYKEG VGKFIGMPES LLYCSLHDPV SPCPAGYVTN KSVSVWSVAG RVEMTVSKFM AIQKALQPDW FQCLSDGEVS CKEATSIKRV RKSVDRSLLF LDNCLRLQEE SEVLQKSVII GVIEGGDVME ERLRSARETA KRPVGGFLLD GFQGNPTTLE ARLRLLSSVT AELPEDKPRL ISGVSRPDEV LECIERGVDL FESFFPYQVT ERGCALTFSF DYQPNPEETL LQQNGTQEEI KCMDQIKKIE TTGCNQEITS FEINLKEKKY QEDFNPLVRG CSCYCCKNHT RAYIHHLLVT NELLAGVLLM MHNFEHYFGF FHYIREALKS DKLAQLKELI HRQAS
Interacts with QTRT1 to form an active queuine tRNA-ribosyltransferase. This enzyme exchanges queuine for the guanine at the wobble position of tRNAs with GU(N) anticodons (tRNA-Asp, -Asn, -His and -Tyr), thereby forming the hypermodified nucleoside queuosine (Q) (7-(((4,5-cis-dihydroxy-2-cyclopenten-1-yl)amino)methyl)-7-deazaguanosine).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Queuine tRNA-ribosyltransferase accessory subunit 2; queuine tRNA-ribosyltransferase domain containing 1; Queuine tRNA-ribosyltransferase domain-containing protein 1
Gene Aliases: QTRT2; QTRTD1
UniProt ID: (Human) Q9H974
Entrez Gene ID: (Human) 79691
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support