Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The QSOX1 gene, also known as Quiescin Q6, is a fusion of two ancient genes: thioredoxin and ERV1. Its expression is induced as fibroblasts begin to exit the proliferative cycle and enter quiescence, suggesting that this gene plays an important role in growth regulation. The QSOX1 protein oxidizes sulfhydryl groups to form disulfide bonds in proteins. QSOX1 expression is induced by hypoxia and appears to protect cells against oxidative stress-induced apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FAD-dependent sulfhydryl oxidase-2; hQSOX; Quiescin Q6; quiescin Q6 sulfhydryl oxidase 1; RP11-502H18.3; rQSOX; SOx-2; Sulfhydryl oxidase 1; testis tissue sperm-binding protein Li 62n; thiol oxidase 1
Gene Aliases: Q6; QSCN6; Qsox; QSOX1; rSOx; Sox-2; Sox2; UNQ2520/PRO6013
UniProt ID: (Human) O00391, (Rat) Q6IUU3
Entrez Gene ID: (Human) 5768, (Rat) 84491
Molecular Function: oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support