Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
R3HDM1 (R3H Domain Containing 1) is a Protein Coding gene. Diseases associated with R3HDM1 include Ichthyosis, Congenital, Autosomal Recessive 11 and Autosomal Recessive Congenital Ichthyosis. Gene Ontology (GO) annotations related to this gene include nucleic acid binding. An important paralog of this gene is R3HDM2. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: R3H domain (binds single-stranded nucleic acids); R3H domain (binds single-stranded nucleic acids) containing; R3H domain 1 (binds single-stranded nucleic acids); R3H domain-containing protein 1
Gene Aliases: KIAA0029; R3HDM; R3HDM1
UniProt ID: (Human) Q15032
Entrez Gene ID: (Human) 23518, (Mouse) 226412
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support