Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive Control: A549 cell lysate, HeLa cell lysate, MCF7 cell lysate, HCT 116 cell lysate, COS-1 cell lysate, NIH/3T3 cell lysate, C6 cell lysate, mouse brain tissue lysate, rat brain tissue lysate.
Subcellular Location: Cytoplasmic vesicle membrane.
Predicted band size: 23 kDa.
RAB10 belongs to the RAS superfamily of small GTPases. RAB proteins localize to exocytic and endocytic compartments and regulate intracellular vesicle trafficking.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-binding protein RAB10; ras-related GTP-binding protein; Ras-related protein Rab-10; ras-related protein rab10
Gene Aliases: Ac1075; AW107754; RAB10
UniProt ID: (Human) P61026, (Mouse) P61027
Entrez Gene ID: (Human) 10890, (Mouse) 19325, (Rat) 50993
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support