Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
MA5-31877 has been tested in indirect ELISA.
Rab11-FIP1 (Rab 11 family-interacting protein 1), also known as Rab-coupling protein (RCP), is a 1283 amino acid Rab 11 effector protein. Rab11-FIP1, by interacting with Rab GTPases, is involved in the endosomal recycling process and may play a role in controlling membrane trafficking along the phagocytic pathway and during phagocytosis. Localized to the recycling endosome, the cytoplasmic membrane and phagosome membranes, Rab11-FIP1 is expressed as five isoforms produced by alternative splicing. As the most highly expressed isoform, isoform two of Rab11-FIP1 is expressed in brain, lung, testis, small intestine, spleen and heart. Isoform two of Rab11-FIP1 also has been found to form a homooligomer and is believed to interact with many Rab GTPases, including Rab 4A, Rab 11A, Rab 11B and Rab 25.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CBBM; hCG_41347; Rab coupling protein; Rab effector protein; Rab-coupling protein; Rab-interacting recycling protein; RAB11 coupling protein; RAB11 family interacting protein 1 (class I); Rab11 family-interacting protein 1; Rab11-FIP1
Gene Aliases: 2010200K21Rik; 4833414G05Rik; NOEL1A; rab11-FIP1; RAB11FIP1; RCP
UniProt ID: (Human) Q6WKZ4, (Mouse) Q9D620
Entrez Gene ID: (Human) 80223, (Mouse) 75767
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support