Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MALVRGAEP AAGPSRWLPT HVQVTVLRAR GLRGKSSGAG STSDAYTVIQ VGREKYSTSV VEKTHGCPEW REECSFELPP GALDGLLRAQ EADAGPAPWA ASSAAACELV LTTMHRSLIG VDKFLGQATV ALDEVFGAGR AQHTQWYKLH SKPGKKEKER GEIEVTIQFT RNNLSAMFDL SMKDKPRSPF SKIRDKMKGK KKYDLESASA ILPSSAIEDP DLGSLGKMGK AKGFFLRNKL RKSSLTQSNT S (1-250 aa encoded by BC035013)
Rab effector involved in protein trafficking from apical recycling endosomes to the apical plasma membrane. Involved in insulin granule exocytosis. May regulate V-ATPase intracellular transport in response to extracellular acidosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gaf-1; Gamma-SNAP-associated factor 1; Phosphoprotein pp75; RAB11 family interacting protein 5 (class I); Rab11 family-interacting protein 5; Rab11-FIP5; Rab11-interacting protein Rip11
Gene Aliases: 9130206P09Rik; C75969; D6Ertd32e; GAF1; KIAA0857; pp75; RAB11FIP5; RIP11
UniProt ID: (Human) Q9BXF6, (Mouse) Q8R361
Entrez Gene ID: (Human) 26056, (Rat) 312502, (Mouse) 52055
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support