Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALVRGAEPA AGPSRWLPTH VQVTVLRARG LRGKSSGAGS TSDAYTVIQV GREKYSTSVV EKTHGCPEWR EECSFELPPG ALDGLLRAQE ADAGPAPWAA SSAAACELVL TTMHRSLIGV DKFLGQATVA LDEVFGAGRA QHTQWYKLHS KPGKKEKERG EIEVTIQFTR NNLSAMFDLS MKDKPRSPFS KIRDKMKGKK KYDLESASAI LPSSAIEDPD LGSLGKMGKA KGFFLRNKLR KSSLTQSNTS LGSDSTLSSA SGSLAYQGPG AELLTRSPSR SSWLSTEGGR DSAQSPKLFT HKRTYSDEAN QMRVAPPRAL LDLQGHLDAA SRSSLCVNGS HIYNEEPQGP VRHRSSISGS LPSSGSLQAV SSRFSEEGPR STDDTWPRGS RSNSSSEAVL GQEELSAQAK VLAPGASHPG EEEGARLPEG KPVQVATPIV ASSEAVAEKE GARKEERKPR MGLFHHHHQG LSRSELGRRS SLGEKGGPIL GASPHHSSSG EGKAKSSWFG LREAKDPTQK PSPHPVKPLS AAPVEGSPDR KQSRSSLSIA LSSGLEKLKT VTSGSIQPVT QAPQAGQMVD TKRLKDSAVL DQSAKYYNLT HDELISLLLQ RERELSQRDE HVQELESYID RLLVRIMETS PTLLQIPPGP PK
Rab effector involved in protein trafficking from apical recycling endosomes to the apical plasma membrane. Involved in insulin granule exocytosis. May regulate V-ATPase intracellular transport in response to extracellular acidosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gaf-1; Gamma-SNAP-associated factor 1; Phosphoprotein pp75; RAB11 family interacting protein 5 (class I); Rab11 family-interacting protein 5; Rab11-FIP5; Rab11-interacting protein Rip11
Gene Aliases: GAF1; KIAA0857; pp75; RAB11FIP5; RIP11
UniProt ID: (Human) Q9BXF6
Entrez Gene ID: (Human) 26056
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support