Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: AWLTEIHEYA QHDVALMLLG NKVDSAHERV VKREDGEKLA KEYGLPFMET SAKTGLNVDL AFTAIAKELK RRSMKAPSEP RFRLHDYVKR EGRGASCC
Members of the RAB protein family, including RAB26, are important regulators of vesicular fusion and trafficking. The RAB family of small G proteins regulates intercellular vesicle trafficking, including exocytosis, endocytosis, and recycling (summary by Seki et al., 2000 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ras-related oncogene protein; Ras-related protein Rab-26
Gene Aliases: RAB26; V46133
UniProt ID: (Human) Q9ULW5
Entrez Gene ID: (Human) 25837
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support