Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
This antibody detects human, mouse, and rat RAB27A in Western blots and recombinant human RAB27A in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human RAB27B is observed.
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
The protein encoded by this gene belongs to the small GTPase superfamily, Rab family. The protein is membrane-bound and may be involved in protein transport and small GTPase mediated signal transduction. Mutations in this gene are associated with Griscelli syndrome type 2. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified. Plays a role in cytotoxic granule exocytosis in lymphocytes. Required for both granule maturation and granule docking and priming at the immunologic synapse.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ashen; GTP-binding protein Ram; low Mr GTP-binding protein; mutant Ras-related protein Rab-27A; Rab-27; Ram p25; Ras-related protein Rab-27A
Gene Aliases: 2210402C08Rik; 2410003M20Rik; 4933437C11Rik; ash; GS2; HsT18676; RAB27; RAB27A; RAM
UniProt ID: (Human) P51159, (Mouse) Q9ERI2, (Rat) P23640
Entrez Gene ID: (Human) 5873, (Mouse) 11891, (Rat) 50645
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support