Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQAPHKEHLY KLLVIGDLGV GKTSIIKRYV HQNFSSHYRA TIGVDFALKV LHWDPETVVR LQLWDIAGQE RFGNMTRVYY REAMGAFIVF DVTRPATFEA VAKWKNDLDS KLSLPNGKPV SVVLLANKCD QGKDVLMNNG LKMDQFCKEH GFVGWFETSA KENINIDEAS RCLVKHILAN ECDLMESIEP DVVKPHLTST KVASCSGCAK S
May be involved in melanosomal transport and docking. Involved in the proper sorting of TYRP1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Melanoma antigen NY-MEL-1; Rab-related GTP-binding protein; Ras-related protein Rab-38
Gene Aliases: NY-MEL-1; RAB38; rrGTPbp
UniProt ID: (Human) P57729
Entrez Gene ID: (Human) 23682
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support