Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MEAIWLYQFR LIVIGDSTVG KSCLIRRFTE GRFAQVSDPT VGVDFFSRLV EIEPGKRIKL QIWDTAGQER FRSITRAYYR NSVGGLLLFD ITNRRSFQNV HEWLEETKVH VQPYQIVFVL VGHKCDLDTQ RQVTRHEAEK LAAAYGMKYI ETSARDAINV EKAFTDLTRD IYELVKRGEI TIQEGWEGVK SGFVPNVVHS SEEVVKSERR CLC
This gene encodes a member of the Rab family of proteins. Rab proteins are small GTPases that are involved in vesicular trafficking.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ras-related protein Rab-39B
Gene Aliases: MRX72; RAB39B; WSMN
UniProt ID: (Human) Q96DA2
Entrez Gene ID: (Human) 116442
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support