Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The RAB6B prodtein seems to have a role in retrograde membrane traffic at the level of the Golgi complex. It may also function in retrograde transport in neuronal cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ras-related protein Rab-6B; small GTP-binding protein; small GTPase RAB6B
Gene Aliases: 9630015C03; AI844641; C330006L04Rik; D9Bwg0185e; RAB6B
UniProt ID: (Human) Q9NRW1, (Mouse) P61294
Entrez Gene ID: (Human) 51560, (Mouse) 270192, (Rat) 363123
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support