Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAKTYDYLF KLLLIGDSGV GKTCLLFRFS EDAFNTTFIS TIGIDFKIRT IELDGKKIKL QIWDTAGQER FRTITTAYYR GAMGIMLVYD ITNEKSFDNI KNWIRNIEEH ASSDVERMIL GNKCDMNDKR QVSKERGEKL AIDYGIKFLE TSAKSSANVE EAFFTLARDI MTKLNRKMND SNSAGAGGPV KITENRSKKT SFFRCSLL (1-207 aa encoded by BC020654)
The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab superfamilies exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. The transport of newly synthesized proteins from the endoplasmic reticulum to various stacks of the Golgi complex and to secretory vesicles involves at each stage the movement of carrier vesicles, a process that appears to involve Rab protein function. Rab 8B (Ras-related protein Rab-8B) is a 207 amino acid lipid anchor that localizes to the cytoplasmic side of the cell membrane and belongs to the Rab family and small GTPase superfamily. Rab 8B may be involved in neurotransmitter release and vesicular trafficking, and is encoded by a gene located on human chromosome Rab 8B and mouse chromosome 9 C.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ38125; RAB-8b protein; RAB44, member RAS oncogene family; Ras-related protein Rab-8B
Gene Aliases: 5930437D16; D330025I23Rik; RAB8B
UniProt ID: (Human) Q92930, (Mouse) P61028
Entrez Gene ID: (Human) 51762, (Mouse) 235442
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support