Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQAIKCVVV GDGAVGKTCL LISYTTNAFP GEYIPTVFDN YSANVMVDSK PVNLGLWDTA GQEDYDRLRP LSYPQTDVFL ICFSLVSPAS YENVRAKWFP EVRHHCPSTP IILVGTKLDL RDDKDTIEKL KEKKLAPITY PQGLALAKEI DSVKYLECSA LTQRGLKTVF DEAIRAVLCP QPTRQQKRAC SLL (1-192 aa encoded by BC001485)
RAC, a small GTPase in the ras family, regulates the formation of membrane ruffles, lamellipodia and filopodia. It is involved in cytoskeletal actin organization and transformation. The p21 activated kinases (PAKs) are direct targets of active Rac and Cdc42 which can induce the assembly of polarized cytoskeletal structures when expressed in fibroblasts. RAC1 and RAC2 are also essential components of NADPH oxidase, the enzyme responsible for generating free radicals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EN-7 protein; GX; p21-Rac2; Protein EN-7; RAS-related C3 botulinum substrate 2; Ras-related C3 botulinum toxin substrate 2; Ras-related C3 botulinum toxin substrate 3 (rho family, small GTP-binding protein Rac2); Small G protein
Gene Aliases: AI323801; AI452260; EN-7; Gx; HSPC022; p21-Rac2; RAC2
UniProt ID: (Human) P15153, (Mouse) Q05144
Entrez Gene ID: (Human) 5880, (Rat) 366957, (Mouse) 19354
Molecular Function:
small GTPase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support