Product References
DNA repair protein FANCD2 has both ubiquitination-dependent and ubiquitination-independent functions during germ cell development.
The Journal of biological chemistry
Zhao S,Huang C,Yang Y,Xu W,Yu Y,Wen C,Cao L,Gao F,Qin Y,Chen ZJ,Guo T,Zhao S
PA5-27195 was used in Immunohistochemistry-immunofluorescence to suggest that FANCD2 has distinct functions that are both dependent on and independent of its ubiquitination during germ cell development.
Wed Mar 01 00:00:00 UTC 2023
LncRNA CTBP1-DT-encoded microprotein DDUP sustains DNA damage response signalling to trigger dual DNA repair mechanisms.
Nucleic acids research
Yu R,Hu Y,Zhang S,Li X,Tang M,Yang M,Wu X,Li Z,Liao X,Xu Y,Li M,Chen S,Qian W,Gong LY,Song L,Li J
PA5-27195 was used in Immunocytochemistry-immunofluorescence to find that DNA damage provoked the association of ribosomes with IRES region in lncRNA CTBP1-DT, which overcame the negative effect of upstream open reading frames (uORFs), and elicited the novel microprotein DNA damage-upregulated protein (DDUP) translation via a cap-independent translation mechanism.
Fri Aug 12 00:00:00 UTC 2022
RAD51 is essential for spermatogenesis and male fertility in mice.
Cell death discovery
Qin J,Huang T,Wang J,Xu L,Dang Q,Xu X,Liu H,Liu Z,Shao C,Zhang X
Published figure using RAD51 polyclonal antibody (Product # PA5-27195) in Immunocytochemistry
Tue Mar 15 00:00:00 UTC 2022
RAD51 is essential for spermatogenesis and male fertility in mice.
Cell death discovery
Qin J,Huang T,Wang J,Xu L,Dang Q,Xu X,Liu H,Liu Z,Shao C,Zhang X
Published figure using RAD51 polyclonal antibody (Product # PA5-27195) in Immunocytochemistry
Tue Mar 15 00:00:00 UTC 2022
RAD51 is essential for spermatogenesis and male fertility in mice.
Cell death discovery
Qin J,Huang T,Wang J,Xu L,Dang Q,Xu X,Liu H,Liu Z,Shao C,Zhang X
Published figure using RAD51 polyclonal antibody (Product # PA5-27195) in Immunocytochemistry
Tue Mar 15 00:00:00 UTC 2022
Complementary anti-cancer pathways triggered by inhibition of sideroflexin 4 in ovarian cancer.
Scientific reports
Tesfay L,Paul BT,Hegde P,Brewer M,Habbani S,Jellison E,Moore T,Wu H,Torti SV,Torti FM
Published figure using RAD51 polyclonal antibody (Product # PA5-27195) in Immunocytochemistry
Sat Nov 19 00:00:00 UTC 2022
MEIOK21 regulates oocyte quantity and quality via modulating meiotic recombination.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Shang Y,Huang J,Li W,Zhang Y,Zhou X,Shao Q,Tan T,Yin S,Zhang L,Wang S
PA5-27195 was used in Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot to indicate critical roles for MEIOK21 in ensuring the number and quality of oocytes in the follicles.
Wed Jun 01 00:00:00 UTC 2022
Selinexor is a novel inhibitor of DNA damage response in Merkel cell carcinoma.
Clinical and experimental dermatology
Moore SA,Narayanan D,Simonette RA,Bartley BR,Doan HQ,Rady PL,Tyring SK
PA5-27195 was used in Gel Electrophoresis to show that Selinexor inhibited DDR protein expression in both MCPyV-positive and MCPyV-negative cells, so addition of Selinexor alone or combined with ICI may be a promising treatment for MCC.
Fri Jul 01 00:00:00 UTC 2022
Cilia regulate meiotic recombination in zebrafish.
Journal of molecular cell biology
Xie H,Wang X,Jin M,Li L,Zhu J,Kang Y,Chen Z,Sun Y,Zhao C
PA5-27195 was used in Immunohistochemistry to reveal a previously undiscovered role for cilia during meiosis and suggests that extracellular signals may regulate meiotic recombination via this particular organelle.
Tue Sep 27 00:00:00 UTC 2022
Gut Microbiota Dysbiosis Accelerates Prostate Cancer Progression Through Increased LPCAT1 Expression and Enhanced DNA Repair Pathways.
Frontiers in oncology
Liu Y,Yang C,Zhang Z,Jiang H
Published figure using RAD51 polyclonal antibody (Product # PA5-27195) in Western Blot
Tue Jul 06 00:00:00 UTC 2021
Gut Microbiota Dysbiosis Accelerates Prostate Cancer Progression Through Increased LPCAT1 Expression and Enhanced DNA Repair Pathways.
Frontiers in oncology
Liu Y,Yang C,Zhang Z,Jiang H
Published figure using RAD51 polyclonal antibody (Product # PA5-27195) in Western Blot
Tue Jul 06 00:00:00 UTC 2021
Gut Microbiota Dysbiosis Accelerates Prostate Cancer Progression Through Increased LPCAT1 Expression and Enhanced DNA Repair Pathways.
Frontiers in oncology
Liu Y,Yang C,Zhang Z,Jiang H
Published figure using RAD51 polyclonal antibody (Product # PA5-27195) in Western Blot
Tue Jul 06 00:00:00 UTC 2021
Chloroxine overrides DNA damage tolerance to restore platinum sensitivity in high-grade serous ovarian cancer.
Cell death & disease
Silva VL,Saxena J,Nicolini F,Hoare JI,Metcalf S,Martin SA,Lockley M
PA5-27195 was used in Immunocytochemistry, Western Blot to investigate the activity of chloroxine as an anticancer agent and its cellular mechanism as well as its synergy with platinum agents in resistant high-grade serous cancer both in vitro and in vivo.
Wed Apr 14 00:00:00 UTC 2021
Chloroxine overrides DNA damage tolerance to restore platinum sensitivity in high-grade serous ovarian cancer.
Cell death & disease
Silva VL,Saxena J,Nicolini F,Hoare JI,Metcalf S,Martin SA,Lockley M
PA5-27195 was used in Immunocytochemistry, Western Blot to investigate the activity of chloroxine as an anticancer agent and its cellular mechanism as well as its synergy with platinum agents in resistant high-grade serous cancer both in vitro and in vivo.
Wed Apr 14 00:00:00 UTC 2021
Chloroxine overrides DNA damage tolerance to restore platinum sensitivity in high-grade serous ovarian cancer.
Cell death & disease
Silva VL,Saxena J,Nicolini F,Hoare JI,Metcalf S,Martin SA,Lockley M
PA5-27195 was used in Immunocytochemistry, Western Blot to investigate the activity of chloroxine as an anticancer agent and its cellular mechanism as well as its synergy with platinum agents in resistant high-grade serous cancer both in vitro and in vivo.
Wed Apr 14 00:00:00 UTC 2021
Chloroxine overrides DNA damage tolerance to restore platinum sensitivity in high-grade serous ovarian cancer.
Cell death & disease
Silva VL,Saxena J,Nicolini F,Hoare JI,Metcalf S,Martin SA,Lockley M
PA5-27195 was used in Immunocytochemistry, Western Blot to investigate the activity of chloroxine as an anticancer agent and its cellular mechanism as well as its synergy with platinum agents in resistant high-grade serous cancer both in vitro and in vivo.
Wed Apr 14 00:00:00 UTC 2021
CHK1 protects oncogenic KRAS-expressing cells from DNA damage and is a target for pancreatic cancer treatment.
Cell reports
Klomp JE,Lee YS,Goodwin CM,Papke B,Klomp JA,Waters AM,Stalnecker CA,DeLiberty JM,Drizyte-Miller K,Yang R,Diehl JN,Yin HH,Pierobon M,Baldelli E,Ryan MB,Li S,Peterson J,Smith AR,Neal JT,McCormick AK,Kuo CJ,Counter CM,Petricoin EF,Cox AD,Bryant KL,Der CJ
PA5-27195 was used in Immunocytochemistry-immunoflourescence to conclude that concurrent DDR suppression enhances the efficacy of ERK and/or autophagy inhibitors in KRAS mutant PDAC.
Tue Nov 30 00:00:00 UTC 2021
A phase I dose-finding, pharmacokinetics and genotyping study of olaparib and lurbinectedin in patients with advanced solid tumors.
Scientific reports
Poveda A,Oaknin A,Romero I,Guerrero-Zotano A,Fariñas-Madrid L,Rodriguez-Freixinos V,Mallol P,Lopez-Reig R,Lopez-Guerrero JA
PA5-27195 was used in Western Blotting to determine recommended, dose-limiting toxicities, and maximum tolerated doses of anti-cancer drugs in ovarian and breast cancer.
Wed Feb 24 00:00:00 UTC 2021
MEIOK21: a new component of meiotic recombination bridges required for spermatogenesis.
Nucleic acids research
Shang Y,Huang T,Liu H,Liu Y,Liang H,Yu X,Li M,Zhai B,Yang X,Wei Y,Wang G,Chen Z,Wang S,Zhang L
PA5-27195 was used in Immunohistochemistry to investigate if MEIOK21 is a component of meiotic recombination bridges and thus required for efficient formation of DMC1/RAD51 foci.
Thu Jul 09 00:00:00 UTC 2020
MEIOK21: a new component of meiotic recombination bridges required for spermatogenesis.
Nucleic acids research
Shang Y,Huang T,Liu H,Liu Y,Liang H,Yu X,Li M,Zhai B,Yang X,Wei Y,Wang G,Chen Z,Wang S,Zhang L
PA5-27195 was used in Immunohistochemistry to investigate if MEIOK21 is a component of meiotic recombination bridges and thus required for efficient formation of DMC1/RAD51 foci.
Thu Jul 09 00:00:00 UTC 2020
The BRCA2-MEILB2-BRME1 complex governs meiotic recombination and impairs the mitotic BRCA2-RAD51 function in cancer cells.
Nature communications
Zhang J,Gurusaran M,Fujiwara Y,Zhang K,Echbarthi M,Vorontsov E,Guo R,Pendlebury DF,Alam I,Livera G,Emmanuelle M,Wang PJ,Nandakumar J,Davies OR,Shibuya H
PA5-27195 was used in Immunocytochemistry-immunoflourescence to identify BRCA2 and MEILB2-associating protein 1 (BRME1), which functions as a stabilizer of MEILB2 by binding to an α-helical N-terminus of MEILB2 and preventing MEILB2 self-association.
Tue Apr 28 00:00:00 UTC 2020
The BRCA2-MEILB2-BRME1 complex governs meiotic recombination and impairs the mitotic BRCA2-RAD51 function in cancer cells.
Nature communications
Zhang J,Gurusaran M,Fujiwara Y,Zhang K,Echbarthi M,Vorontsov E,Guo R,Pendlebury DF,Alam I,Livera G,Emmanuelle M,Wang PJ,Nandakumar J,Davies OR,Shibuya H
PA5-27195 was used in Immunocytochemistry-immunoflourescence to identify BRCA2 and MEILB2-associating protein 1 (BRME1), which functions as a stabilizer of MEILB2 by binding to an α-helical N-terminus of MEILB2 and preventing MEILB2 self-association.
Tue Apr 28 00:00:00 UTC 2020
The BRCA2-MEILB2-BRME1 complex governs meiotic recombination and impairs the mitotic BRCA2-RAD51 function in cancer cells.
Nature communications
Zhang J,Gurusaran M,Fujiwara Y,Zhang K,Echbarthi M,Vorontsov E,Guo R,Pendlebury DF,Alam I,Livera G,Emmanuelle M,Wang PJ,Nandakumar J,Davies OR,Shibuya H
PA5-27195 was used in Immunocytochemistry-immunoflourescence to identify BRCA2 and MEILB2-associating protein 1 (BRME1), which functions as a stabilizer of MEILB2 by binding to an α-helical N-terminus of MEILB2 and preventing MEILB2 self-association.
Tue Apr 28 00:00:00 UTC 2020
The histone modification reader ZCWPW1 links histone methylation to PRDM9-induced double-strand break repair.
eLife
Huang T,Yuan S,Gao L,Li M,Yu X,Zhan J,Yin Y,Liu C,Zhang C,Lu G,Li W,Liu J,Chen ZJ,Liu H
PA5-27195 was used in Immunocytochemistry to demonstrate the function of ZCWPW1 that acts as part of the selection system for epigenetics-based recombination hotspots in mammals.
Wed May 06 00:00:00 UTC 2020
SCRE serves as a unique synaptonemal complex fastener and is essential for progression of meiosis prophase I in mice.
Nucleic acids research
Liu H,Huang T,Li M,Li M,Zhang C,Jiang J,Yu X,Yin Y,Zhang F,Lu G,Luo MC,Zhang LR,Li J,Liu K,Chen ZJ
PA5-27195 was used in Immunocytochemistry-immunoflourescence to identify a novel protein, synaptonemal complex reinforcing element as a key molecule in maintaining the integrity of synaptonemal complex during meiosis prophase 1 in mice.
Thu Jun 20 00:00:00 UTC 2019
A meiosis-specific BRCA2 binding protein recruits recombinases to DNA double-strand breaks to ensure homologous recombination.
Nature communications
Zhang J,Fujiwara Y,Yamamoto S,Shibuya H
PA5-27195 was used in Western Blotting to study the role of BRCA2 in the repair mechanism of double stranded breaks.
Wed Feb 13 00:00:00 UTC 2019
A meiosis-specific BRCA2 binding protein recruits recombinases to DNA double-strand breaks to ensure homologous recombination.
Nature communications
Zhang J,Fujiwara Y,Yamamoto S,Shibuya H
PA5-27195 was used in Western Blotting to study the role of BRCA2 in the repair mechanism of double stranded breaks.
Wed Feb 13 00:00:00 UTC 2019
Drugging DNA repair to target T-ALL cells.
Leukemia & lymphoma
Dasgupta Y,Golovine K,Nieborowska-Skorska M,Luo L,Matlawska-Wasowska K,Mullighan CG,Skorski T
Published figure using RAD51 polyclonal antibody (Product # PA5-27195) in Immunocytochemistry
Sun Jul 01 00:00:00 UTC 2018
Rgf1p (Rho1p GEF) is required for double-strand break repair in fission yeast.
Nucleic acids research
Manjón E,Edreira T,Muñoz S,Sánchez Y
PA5-27195 was used in Immunohistochemistry-immunofluorescence to explore the sensitivity of a deletion mutation in the Rho1p exchange factor Rgf1p to the DNA break/inducing antibiotic phleomycin.
Fri May 19 00:00:00 UTC 2017