Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-17163 targets RAG1 in indirect ELISA and WB applications and shows reactivity with Human samples.
The MA5-17163 immunogen is purified recombinant fragment of human RAG1 (amino acids: 818-868) expressed in E. Coli.
MA5-17163 detects RAG1 which has a predicted molecular weight of approximately 119kDa.
The protein encoded by this gene is involved in activation of immunoglobulin V-D-J recombination. The encoded protein is involved in recognition of the DNA substrate, but stable binding and cleavage activity also requires RAG2. Defects in this gene can be the cause of several diseases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC43321; RAG-1; RAG-1 Antibody; recombination activating gene 1 Antibody; recombination activating protein 1; RING finger protein 74; RNF74 Antibody; V(D)J recombination-activating protein 1
Gene Aliases: RAG-1; RAG1; RNF74
UniProt ID: (Human) P15918
Entrez Gene ID: (Human) 5896
Molecular Function:
DNA metabolism protein
endodeoxyribonuclease
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support