Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Specificity: 2A11 is specific for the V-domain of human advanced glycosylation end product-specific receptor. It recognises both the glycosylated and non-glycosylated forms.
Able to phosphorylate several exogenous substrates and to undergo autophosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Advanced glycosylation end product-specific receptor; RAGE isoform NtRAGE-delta; RAGE isoform sRAGE-delta; RAGE-4 ORF3; receptor for advanced glycation end-products variant 20; Receptor for advanced glycosylation end products; sRAGE
Gene Aliases: AGER; RAGE; SCARJ1
UniProt ID: (Human) Q15109
Entrez Gene ID: (Human) 177
Molecular Function: cell adhesion molecule immunoglobulin superfamily cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support