Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
Immunogen sequence is different from the related mouse and rat sequences by six amino acids.
Able to phosphorylate several exogenous substrates and to undergo autophosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: advanced glycation end-products receptor; Advanced glycosylation end product-specific receptor; advanced glycosylation end product-specific receptor variant 2; advanced glycosylation end product-specific receptor variant 3; advanced glycosylation end product-specific receptor variant 4; advanced glycosylation end product-specific receptor variant 5; RAGE isoform NtRAGE-delta; RAGE isoform sRAGE-delta; RAGE-4 ORF3; receptor for advanced glycation end-products variant 20; Receptor for advanced glycosylation end products; sRAGE
Gene Aliases: AGER; RAGE; SCARJ1
UniProt ID: (Human) Q15109, (Mouse) Q62151
Entrez Gene ID: (Human) 177, (Rat) 81722, (Mouse) 11596
Molecular Function:
cell adhesion molecule
immunoglobulin superfamily cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support