Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
MOK is able to phosphorylate several exogenous substrates and to undergo autophosphorylation. It negatively regulates cilium length in a cAMP and mTORC1 signaling-dependent manner.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAPK/MAK/MRK overlapping kinase; MOK protein kinase; RAGE-1; renal cell carcinoma antigen; Renal tumor antigen 1; soluble RAGE
Gene Aliases: MOK; RAGE; RAGE-1; RAGE1; STK30
UniProt ID: (Human) Q9UQ07
Entrez Gene ID: (Human) 5891
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support