Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of total AS250.
Catalytic subunit of the heterodimeric RalGAP2 complex which acts as a GTPase activator for the Ras-like small GTPases RALA and RALB.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 250 kDa substrate of Akt; akt substrate AS250; AS250; P220; Ral GTPase activating protein, alpha subunit 2 (catalytic); ral GTPase-activating protein alpha subunit 2; Ral GTPase-activating protein subunit alpha-2; RapGAPalpha2
Gene Aliases: A230067G21Rik; AS250; bA287B20.1; BC053994; C20orf74; dJ1049G11; dJ1049G11.4; KIAA1272; p220; pp250; RALGAPA2; RGC2
UniProt ID: (Human) Q2PPJ7, (Mouse) A3KGS3
Entrez Gene ID: (Human) 57186, (Mouse) 241694
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support