Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 5
Non-catalytic subunit of the heterodimeric RalGAP1 and RalGAP2 complexes which act as GTPase activators for the Ras-like small GTPases RALA and RALB.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781M2411; p170; p170 {ECO:0000303|PubMed:19520869}; Ral GTPase activating protein, beta subunit (non-catalytic); ral GTPase-activating protein beta subunit; Ral GTPase-activating protein subunit beta; ralgapb {ECO:0000303|PubMed:19520869}
Gene Aliases: 9330195P16; AI507211; B230339M05Rik; KIAA1219; mKIAA1219; p170; RALGAPB; RalGAPbeta; RGD1306861
UniProt ID: (Human) Q86X10
Entrez Gene ID: (Human) 57148, (Mouse) 228850, (Rat) 362257
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support