Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat
Non-catalytic subunit of the heterodimeric RalGAP1 and RalGAP2 complexes which act as GTPase activators for the Ras-like small GTPases RALA and RALB.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781M2411; p170; Ral GTPase activating protein, beta subunit (non-catalytic); ral GTPase-activating protein beta subunit; Ral GTPase-activating protein subunit beta
Gene Aliases: 9330195P16; AI507211; B230339M05Rik; KIAA1219; mKIAA1219; p170; RALGAPB; RalGAPbeta
UniProt ID: (Human) Q86X10
Entrez Gene ID: (Human) 57148, (Mouse) 228850
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support