Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LKIEPGTSTP RSAASREDLV GPEVGASPQS GRKSVAAEGA LLPQTPPSPR NLIPHGHRKC HSLGYNFIHK MNTAEFKSAT FPNAGPRHLL DDSVMEPHAP SRGQA
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ral GEF with PH domain and SH3-binding motif 2; Ral-A exchange factor RalGPS2; RalA exchange factor RalGPS2; Ras-specific guanine nucleotide-releasing factor RalGPS2
Gene Aliases: dJ595C2.1; RALGPS2
UniProt ID: (Human) Q86X27
Entrez Gene ID: (Human) 55103
Molecular Function: guanyl-nucleotide exchange factor scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support