Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YPGSPSLSP RHGPSSSHMH NTGADSPSCS NGVASTKSKQ NHSKYPAPSS SSSSSSSSSS SSPSSVNYSE SNSTDSTKSQ HHSSTSNQET SDSEVEMEAE HYPNGVLGSM STRIVNGAYK HEDLQTDESS MDDRHPR (362-497 aa encoded by BC099917 )
The RAN binding protein 10 (RANBP10) is a cytoplasmic guanine nucleotide exchange factor that shares high sequence similarity to the MET-interacting protein RANBP9. RANBP10 is expressed in the cytoplasm of mature megakaryocytes (MKs) and platelets where it localizes to polymerized noncentrosomal microtubules. RANBP10 possesses guanine nucleotide exchange factor activity towards RAN, a GTPase involved in nucleocytoplasmic traffic, spindle formation at mitosis, and nuclear envelope assembly during telophase, suggesting that RANBP10 may also be involved in these activities. RANBP10 depletion in vitro caused the disturbance of polymerized filaments in MKs and platelets in RANBP10-null mice exhibited disorders in microtubule filament numbers and localization, indicating that RANBP10 may play a significant role in maintaining microtubule dynamics.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ran-binding protein 10; RanBP10; rangef
Gene Aliases: 4432417N03Rik; KIAA1464; RANBP10; RGD1311587
UniProt ID: (Human) Q6VN20, (Mouse) Q6VN19
Entrez Gene ID: (Human) 57610, (Mouse) 74334, (Rat) 361396
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support