Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-4995 detects RanBP2 from human samples.
PA1-4995 has been successfully used in Western blot, immunoprecimmunoprecipitationitation, and immunohistochemistry applications.
The PA1-4995 immunogen is synthetic peptide corresponding to residues SKAPKSGFEGMFTKKE, repeated 3 times.
RanBP2 interacts with the small GTP-binding protein Ran and forms a large multiprotein complex involved in many biological processes such as mRNA processing, nuclear transport, cell cycle control and postmitotic nuclear reassembly. The protein is identified by western blotting as an apparent molecular mass of 360 kD. It is involved in nucleation of microtubule and control of cell growth by interacting with many protein factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 358 kDa nucleoporin; acute necrotizing encephalopathy 1 (autosomal dominant); E3 SUMO-protein ligase RanBP2; Nuclear pore complex protein Nup358; nucleoporin 358; Nucleoporin Nup358; P270; Ran-binding protein 2; RanBP2; RBP2; transformation-related protein 2
Gene Aliases: A430087B05Rik; ADANE; AI256741; ANE1; IIAE3; NUP358; RANBP2; RGD1560047; TRP1; TRP2
UniProt ID: (Human) P49792, (Mouse) Q9ERU9
Entrez Gene ID: (Human) 5903, (Rat) 294429, (Mouse) 19386
Molecular Function: scaffold/adaptor protein chaperone transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support