Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detects endogenous protein at a molecular weight of 125 kDa.
Purity is >95% by SDS-PAGE.
RANBP6 (Ran-binding protein 6) belongs to the importin beta family and may function in nuclear protein import as a nuclear transport receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ran BP6; Ran-binding protein 6; Ran-BP6; Ran-GTP binding protein; RanBP6
Gene Aliases: C630001B19; RANBP6
UniProt ID: (Human) O60518, (Mouse) Q8BIV3
Entrez Gene ID: (Human) 26953, (Mouse) 240614, (Rat) 309326
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support