Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MASNSSSCPT PGGGHLNGYP VPPYAFFFPP MLGGLSPPGA LTTLQHQLPV SGYSTPSPAT IETQSSSSEE IVPSPPSPPP LPRIYKPCFV CQDKSSGYHY GVSACEGCKG FFRRSIQKNM VYTCHRDKNC IINKVTRNRC QYCRLQKCFE VGMSKESVRN DRNKKKKEVP KPECSESYTL TPEVGELIEK VRKAHQETFP ALCQLGKYTT NNSSEQRVSL DIDLWDKFSE LSTKCIIKTV EFAKQLPGFT TLTIADQITL LKAACLDILI LRICTRYTPE QDTMTFSDGL TLNRTQMHNA GFGPLTDLVF AFANQLLPLE MDDAETGLLS AICLICGDRQ DLEQPDRVDM LQEPLLEALK VYVRKRRPSR PHMFPKMLMK ITDLRSISAK GAERVITLKM EIPGSMPPLI QEMLENSEGL DTLSGQPGGG GRDGGGLAPP PGSCSPSLSP SSNRSSPATH SP
RARA is a receptor for retinoic acid. Retinoic acid receptors bind as heterodimers to their target response elements in response to their ligands, all-trans or 9-cis retinoic acid, and regulate gene expression in various biological processes. The RXR/RAR heterodimers bind to the retinoic acid response elements (RARE) composed of tandem 5'-AGGTCA-3' sites known as DR1-DR5. In the absence of ligand, the RXR-RAR heterodimers associate with a multiprotein complex containing transcription corepressors that induce histone acetylation, chromatin condensation and transcriptional suppression. On ligand binding, the corepressors dissociate from the receptors and associate with the coactivators leading to transcriptional activation. RARA plays an essential role in the regulation of retinoic acid-induced germ cell development during spermatogenesis. Has a role in the survival of early spermatocytes at the beginning prophase of meiosis. In Sertoli cells, may promote the survival and development of early meiotic prophase spermatocytes. In concert with RARG, required for skeletal growth, matrix homeostasis and growth plate function. Regulates expression of target genes in a ligand-dependent manner by recruiting chromatin complexes containing KMT2E/MLL5. Mediates retinoic acid-induced granulopoiesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nuclear receptor subfamily 1 group B member 1; nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form; RAR Alpha; RAR-alpha; retinoic acid; retinoic acid nuclear receptor alpha variant 1; retinoic acid nuclear receptor alpha variant 2; retinoic acid receptor; Retinoic acid receptor alpha; retinoic acid receptor, alpha polypeptide; RRA
Gene Aliases: NR1B1; RAR; RARA
UniProt ID: (Human) P10276
Entrez Gene ID: (Human) 5914
Molecular Function:
C4 zinc finger nuclear receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support