Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MATNKERLF AAGALGPGSG YPGAGFPFAF PGALRGSPPF EMLSPSFRGL GQPDLPKEMA SLSVETQSTS SEEMVPSSPS PPPPPRVYKP CFVCNDKSSG YHYGVSSCEG CKGFFRRSIQ KNMVYTCHRD KNCIINKVTR NRCQYCRLQK CFEVGMSKEA VRNDRNKKKK EVKEEGSPDS YELSPQLEEL ITKVSKAHQE TFPSLCQLGK YTTNSSADHR VQLDLGLWDK FSELATKCII KIVEFAKRLP GFTGLSIADQ ITLLKAACLD ILMLRICTRY TPEQDTMTFS DGLTLNRTQM H (1-300 aa encoded by BC019098)
Retinoic acid, a metabolite of vitamin A, is necessary for normal organogenesis but acts as a teratogen at high levels during embryonic and fetal development. Retinoic acid functions through its interaction with the nuclear protein, retinoic acid receptor (RAR). RAR belongs to the steroid and thyroid hormone superfamily of nuclear receptor proteins which exert their effects by binding to specific DNA response elements, thus regulating gene expression in target cells. RAR exists as three major subtypes: alpha, beta and gamma. Two main isoforms of RAR gamma exist: RAR gamma-1 has been found to be predominantly expressed in skin, while RAR gamma-2 has been found early in embryogenesis and in embryonal carcinoma cells. RAR gamma is also known as RARD. The corresponding gene for the retinoic acid receptor gamma 1 is NR1B3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nuclear receptor subfamily 1 group B member 3; RAR gamma 2; RAR Gamma-1; RAR-gamma; retinoic acid; retinoic acid nuclear receptor gamma variant 1; retinoic acid nuclear receptor gamma variant 2; retinoic acid receptor; Retinoic acid receptor gamma; retinoic acid receptor gamma 4; retinoic acid receptor, gamma
Gene Aliases: NR1B3; RAR-gamma; RARC; RARD; RARG; RARgamma2
UniProt ID: (Human) P13631, (Mouse) P18911
Entrez Gene ID: (Human) 5916, (Mouse) 19411, (Rat) 685072
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support